2,2',3,5,6-Pentachlorobiphenyl (PCB No. 93)

Leverantör: LGC Standards PROMOCHEM

CIL-PCB-93-C
PROOCIL-PCB-93-CEA 0 SEK
PROOCIL-PCB-93-C
2,2',3,5,6-Pentachlorobiphenyl (PCB No. 93)
2,2',3,5,6-Pentachlorobiphenyl (PCB No. 93)
Formel: C₁₂H₅Cl₅
MW: 326.44 g/mol
CAS nummer: 73575-56-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR