2,2',3,3',4,4',6,6'-Octachlorobiphenyl (PCB No. 197)

Leverantör: LGC Standards PROMOCHEM

CIL-PCB-197-C
PROOCIL-PCB-197-CEA 0 SEK
PROOCIL-PCB-197-C
2,2',3,3',4,4',6,6'-Octachlorobiphenyl (PCB No. 197)
2,2',3,3',4,4',6,6'-Octachlorobiphenyl (PCB No. 197)
Formel: C₁₂H₂Cl₈
MW: 429.77 g/mol
CAS nummer: 33091-17-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR