2,2',3,3',4,5',6'-Heptachlorobiphenyl (PCB No. 177)

Leverantör: LGC Standards PROMOCHEM

CIL-PCB-177-C
PROOCIL-PCB-177-CEA 0 SEK
PROOCIL-PCB-177-C
2,2',3,3',4,5',6'-Heptachlorobiphenyl (PCB No. 177)
2,2',3,3',4,5',6'-Heptachlorobiphenyl (PCB No. 177)
Formel: C₁₂H₃Cl₇
MW: 395.32 g/mol
CAS nummer: 52663-70-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR