2,2',3,4,5,6-Hexachlorobiphenyl (PCB No. 142)

Leverantör: LGC Standards PROMOCHEM

CIL-PCB-142-C
PROOCIL-PCB-142-CEA 0 SEK
PROOCIL-PCB-142-C
2,2',3,4,5,6-Hexachlorobiphenyl (PCB No. 142)
2,2',3,4,5,6-Hexachlorobiphenyl (PCB No. 142)
Formel: (C₆H₄Cl)C₆Cl₅
MW: 360.88 g/mol
CAS nummer: 41411-61-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR