2,5-Dichlorobiphenyl-[13C12] (PCB No. 9) 40 µg/ml i n-nonan (99% 13C)

Leverantör: LGC Standards PROMOCHEM

CILEC-4165-1.2 CIL-EC-4165-3
PROOCILEC-4165-1.2EA 0 SEK
PROOCILEC-4165-1.2 PROOCIL-EC-4165-3
2,5-Dichlorobiphenyl-[13C12] (PCB No. 9) 40 µg/ml i n-nonan (99% 13C)
Standarder Organic Standards

Order Now

SPECIFIKATIONER

Benämning 2,5-Dichlorobiphenyl-[13C12](PCB No. 9) 2,5-Dichlorobiphenyl-[13C12] (PCB No. 9)
Environmentally Preferable
CAS-nr 25569-80-6 25569-80-6
Formel C₁₂H₈Cl₂ C₁₂H₈Cl₂

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR