Benzylbenzoat

Leverantör: LGC Standards PROMOCHEM
Varning

Synonyms: Benzoic acid benzyl ester

BP426
PROOBP426EA 0 SEK
PROOBP426
Benzylbenzoat
Benzylbenzoat
Formel: C₁₄H₁₂O₂
MW: 212.25 g/mol
Kokpunkt: 323
Smältpunkt: 17…20 °C
Densitet: 1.118 g/cm³ (20 °C)
Flampunkt: 158 °C (closed cup)
Storage Temperature: Omgivande
MDL Number: MFCD00003075
CAS nummer: 120-51-4
EINECS: 204-402-9
UN: 3082
ADR: 9 III
Merck Index: 13,01129

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR