Certified reference material, soya-maize oil blend, BCR®
Leverantör: LGC Standards PROMOCHEM
BCR-162R
PROOBCR-162REA 0 SEK
PROOBCR-162R
Certified reference material, soya-maize oil blend, BCR®
Standarder Livsmedelsstandarder
A unit contains approximately 5,5 gm soya-maize oil blend in 10 ml amber ampoule.
  • Fatty acid analysis
Certifikat: IRMM certified.
Observera: Store in Freezer below –20 °C.