Cobalt spin filter plates, 96 well, HisPur™
Leverantör: Thermo Fisher Scientific
90095
PIER90095EA 8750 SEK
PIER90095
Cobalt spin filter plates, 96 well, HisPur™
Plattor Filterplattor
HisPur™ Cobalt spin plates are convenient, ready-to-use, pre-dispensed filter plates for the efficient purification of polyhistidine-tagged proteins from bacterial, mammalian and baculovirus-infected cells. These spin plates are ideal for simultaneous processing of multiple small (1 mg/well binding capacity) samples.
  • Obtain more than 10 mg of pure His-tagged protein per ml of resin without optimising imidazole washing conditions
  • Cobalt-chelate coordination core binds fewer host protein contaminants, resulting in lower background than nickel resins
  • No metal
More Product Information