UV exposure screen for myECL™ imager, Pierce
Leverantör: Thermo Fisher Scientific
62244
PIER62244EA 11450 SEK
PIER62244
UV exposure screen for myECL™ imager, Pierce
Fotodokumentationssystem
This clear plastic sample screen is used to cover the transilluminator on the myECL™ imager. It is a replacement for the accessory used on the transilluminator for acquiring images in the UV mode.