N,N-Dimetylformamid ≥99.5%, Sequencing grade, Pierce™

Leverantör: Thermo Fisher Scientific
Fara

Synonyms: DMF

20673
PIER20673EA 1275 SEK
PIER20673
N,N-Dimetylformamid ≥99.5%, Sequencing grade, Pierce™
N,N-Dimetylformamid

Thermo Scientific Pierce Dimethylformamide (DMF) is a sequencing-grade preparation with high purity and consistent absorptivity properties for use in HPLC and mass spectrometry methods.


  • Highly purified and tested to ensure reliable performance in HPLC, mass spectrometry and sequencing applications
  • Packaged in solvent-rinsed amber glass bottles and sealed with PTFE-lined fluorocarbon caps


This sequencing-grade dimethylformamide is specially purified by a proprietary method and tested to ensure lot-to-lot consistency with a low UV absorbance to provide you with the most sensitive detection across all wavelengths. DMF is packaged in solvent-rinsed amber glass bottles and sealed with PTFE-lined fluorocarbon caps for ultimate protection.

Pierce sequencing-grade DMF is specially purified and tested to the highest specifications to ensure the integrity of your data, maximize sensitivity in your assay and to prolong the life of your equipment.


Observera: For Research Use Only. Not for use in diagnostic procedures.

Formel: HCON(CH₃)₂
MW: 73.09 g/mol
Kokpunkt: 153
Smältpunkt: –61 °C
Densitet: 0.949 g/cm³ (20 °C)
Flampunkt: 57.5 °C (closed cup)
Storage Temperature: Omgivande
MDL Number: MFCD00003284
CAS nummer: 68-12-2
EINECS: 200-679-5
UN: 2265
ADR: 3 III
REACH: 01-2119475605-32
Merck Index: 13,03269
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=1ac5c5e9758b6c40ccb0cf127b6a6e1c6d608c64&vid=9a5d4e579e195193602c20d3cb39c89ac9c92cbf&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR