Compensated jacket calorimeter, Model 6100
Leverantör: PARR
6100EF
PARR6100EFEA 0 SEK
PARR6100EF
Compensated jacket calorimeter, Model 6100
Kalorimetrar
The 6100 calorimeter is intended for the user who wants a modern calorimeter with the convenient automatic features provided in the 6200 calorimeter and whose precision requirements can be met with a static system without isoperibol control. or, for users whose workload is small or intermittent, making it preferable to purchase a lower cost model.
  • Traditional design calorimeter with removable oxygen bomb and bucket
  • High speed calorimeter capable of performing up to seven tests per hour when equipped with two bombs and buckets
  • Compact calorimeter requiring
More Product Information