GridION™ DNA/RNA Benchtop Nanopore Sequencer
Leverantör: Oxford Nanopore Technologies
SUPP001 GRD-MK1CAPX SUPP006 BASIC12M-G BASIC24M-G BASIC36M-G
OXNTSUPP001EA 0 SEK
OXNTSUPP001 OXNTGRD-MK1CAPX OXNTSUPP006 OXNTBASIC12M-G OXNTBASIC24M-G OXNTBASIC36M-G
GridION™ DNA/RNA Benchtop Nanopore Sequencer
Next Generation Sequencing Instruments
GridION™ - a self-contained, easily deployable DNA/RNA benchtop nanopore sequencer.
  • Flexible - run up to five independently addressable flow cells
  • Integrated compute - powerful onboard data processing and analysis, minimizing IT requirements
  • Compact - small footprint, suitable for any lab
  • Real-time data analysis - rapid results with base modification data as standard
  • Any read length - from short to ultra-long (longest >4 Mb)
A flexible, self-contained, benchtop nanopore sequencer, running up to five MinION™ Flow Cells that can respond to the needs
More Product Information
Order Now
SPECIFIKATIONER
Environmentally Preferable
Please note that Oxford Nanopore Technologies requires the acceptance of their Terms & Conditions prior to product order placement. If you select product(s) to order, the Terms and Conditions will be displayed in the shopping cart for your review and acceptance.