Dropping pipettes, Nalgene®
Leverantör: Thermo Fisher Scientific
6219-0068
NALG6219-0068EA 695 SEK
NALG6219-0068
Dropping pipettes, Nalgene®
Pipetter Pasteurpipetter
LDPE. Moulded in one piece. Non wetting material, tapered tip, and bellows-type bulb provide excellent drop formation.
  • Delivers 30 to 35 drops per ml
  • Safe and economical
  • Easy to clean for repeatable performance
  • Chemically compatible with most acids, bases and alcohols
Order Now
SPECIFIKATIONER
Environmentally Preferable Low Manufacturing Impact
Miljöcertificering IRECS; ISO 14001 (Certificate Registration No. EMS 752603)