6-Bromo-2-naphthol

Leverantör: Molekula

Synonyms: 6-Bromo-2-hydroxynaphthalene, 2-Bromo-6-hydroxynaphthalene

M54071591
MOLEM54071591EA 0 SEK
MOLEM54071591
6-Bromo-2-naphthol
6-Bromo-2-naphthol
Formel: C₁₀H₇BrO
MW: 223.07 g/mol
Kokpunkt: 200…205 °C (20 mmHg)
Smältpunkt: 128
MDL Number: MFCD00004081
CAS nummer: 15231-91-1
EINECS: 239-279-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR