2-Nitrothiophene

Leverantör: Molekula

M50354243
MOLEM50354243EA 0 SEK
MOLEM50354243
2-Nitrothiophene
2-Nitrothiophene
Formel: C₄H₃NO₂S
MW: 129.14 g/mol
Kokpunkt: 224
Smältpunkt: 46.5
Densitet: 1.3644 g/cm³ (42.6 °C)
Storage Temperature: Omgivande
MDL Number: MFCD00005425
CAS nummer: 609-40-5
EINECS: 210-190-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR