1,1'-Carbonyldiimidazole

Leverantör: Molekula
Fara

Synonyms: Di(1H-imidazol-1-yl)methanone, CDI Crosslinker, 1,1'-Carbonylbis(imidazole), 1,1'-Carbonylbis-1H-imidazole, N,N'-Carbonyldiimidazole, CDI

M10944806
MOLEM10944806EA 0 SEK
MOLEM10944806
1,1'-Carbonyldiimidazole
1,1'-Carbonyldiimidazole
Formel: C₇H₆N₄O
MW: 162.15 g/mol
Smältpunkt: 111…116 °C
Storage Temperature: Omgivande
MDL Number: MFCD00005286
CAS nummer: 530-62-1
EINECS: 208-488-9
UN: 3263
ADR: 8 II
Merck Index: 14,01819

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR