D (-)-Mannitol

Leverantör: Molekula

Synonyms: Mannitol, (2R,3R,4R,5R)-Hexane-1,2,3,4,5,6-hexaol

71640365-5KG
MOLE71640365-5KGEA 0 SEK
MOLE71640365-5KG
D (-)-Mannitol
D (-)-Mannitol
Formel: HOH₂C(CH(OH))₄CH₂OH
MW: 182.17 g/mol
Kokpunkt: 290…295 °C (4 hPa)
Smältpunkt: 164…169 °C
Densitet: 1.49 g/cm³ (20 °C)
Storage Temperature: Omgivande
MDL Number: MFCD00064287
CAS nummer: 69-65-8
EINECS: 200-711-8
Merck Index: 13,05769
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=163f556dca45086e05ff490ddc8eeb65d04aea05&vid=408e3f5566e309064ce68988cdd160305c5b8c0a&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR