2-Chloroethylammonium chloride

Leverantör: Molekula

Synonyms: 2-Chloroethylamine hydrochloride, 2-Chloroethylamine HCl, 2-Chloroethanamine hydrochloride

13713550-500G
MOLE13713550-500GEA 0 SEK
MOLE13713550-500G
2-Chloroethylammonium chloride
2-Chloroethylammonium chloride
Formel: C₂H₇Cl₂N
MW: 115.99 g/mol
MDL Number: MFCD00012887
CAS nummer: 870-24-6
EINECS: 212-793-2
UN: 3261
ADR: 8 III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR