Benzyltrimethylammonium chloride

Leverantör: Molekula
Varning

Synonyms: N,N,N-Trimethyl-1-phenylmethanaminium chloride

10717982-1KG
MOLE10717982-1KGEA 0 SEK
MOLE10717982-1KG
Benzyltrimethylammonium chloride
Benzyltrimethylammonium chloride
Formel: C₁₀H₁₆ClN
MW: 185.7 g/mol
Smältpunkt: 238
Densitet: 1.07 g/cm³ (20 °C)
Storage Temperature: Omgivande
MDL Number: MFCD00011782
CAS nummer: 56-93-9
EINECS: 200-300-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR