Capillary dispenser, VITREX®
Leverantör: Vitrex Medical A/S
569013
MODU569013EA 445 SEK
MODU569013
Capillary dispenser, VITREX®
Pipetter Mikrokapillärer
This capillary dispenser is a unique liquid handling tool, developed to empty capillary tubes for blood or other liquids.
  • Two step plunger operation facilitates super blow-out from the tip
  • Its unique tip cone is designed to accept most VITREX capillary tubes
  • At a length of 130 mm this pipette is the optimum practical size for blood collecting
  • Fully autoclavable at 121 °C & 15 psi
This dispenser is specially designed for use with Diagnostic Kits.