Electronic earplug, Peltor™
Leverantör: 3M
EEP-100EU
MMMAEEP-100EUEA 4575 SEK
MMMAEEP-100EU
Electronic earplug, Peltor™
Öronproppar Öronproppar, återanvändbara
Peltor™ Electronic Earplugs have an intuitive one-button operation that turns the earplugs on, off, and selects volume settings. They’re powered by Li-ion batteries, which can be charged via a micro B USB charging port on the pocket-sized storage case, incorporating an easily visible charging status indicator.
  • Protect hearing from sudden loud (impulse) noises
  • Enables ambient sounds below 82 dB to still be heard
  • Supports easy communication with those nearby
  • Li-ion batteries provide up to 16 hours of continuous operation
  • Multiple types and sizes of
More Product Information