Safety stripe tape, 5702
Leverantör: 3M
5702
MMMA5702EA 970 SEK
MMMA5702
Safety stripe tape, 5702
Tejp
This tape is ideal for marking lanes, hazardous areas, protruding equipment, and low hanging objects in factories, warehouses, and hospitals. Vivid, high-visibility yellow and black diagonal stripes that lock into the vinyl alert people about hazardous conditions.
  • Applies instantly and is ready to use immediately
  • Yellow and black stripes are locked into the vinyl for permanent high visibility
  • Stretches and conforms to curved surfaces and around corners for a tight fit
  • Resists abrasion, scuffing, moisture, and weathering for long service life
  • Used to
More Product Information