Multiscreen® HTS IP Filter Plate, Hydrophobic PVDF Membrane
Leverantör: Merck
M8IPS4510
MILFM8IPS4510EA 13925 SEK
MILFM8IPS4510
Multiscreen® HTS IP Filter Plate, Hydrophobic PVDF Membrane
Plattor Filterplattor
MultiScreenHTS plates are specifically developed for high-throughput use with automated work stations. Rigid sidewalls improve handling and ample surfaces are provided for bar code labels.
  • Binding type: High binding surface
  • Shipping condition: Ambient
  • Filtration area: 0,28 cm²
  • Material: Acrylic
  • Flat bottom wells
Wells are individually sealed to prevent incubation crosstalk and the underdrain is removable for access to the filters.