Autosampler vials, 3,8 ml
Leverantör: MICROSOLV
9534S-0CV
MICS9534S-0CVEA 3475 SEK
MICS9534S-0CV
Autosampler vials, 3,8 ml
Vialer
These vials made of TYPE 1 borosilicate glass. The unique center drain offers maximum recovery of compounds placed in these vials for storage or sampling.
  • Sturdy with a 'trumpet' bottom
  • Ultra low dead volume for better recovery with center drain
  • Suitable for use with Waters autosamplers