Masterflex® Ismatec® Pump Tubing, 2-Stop Microbore, Tygon® LMT-55, Avantor®
Leverantör: Avantor Fluid Handling
MFLX97618-09EA 895 SEK
MFLX97618-09 MFLX97618-10 MFLX97618-14 MFLX97618-16 MFLX97618-18 MFLX97618-22 MFLX97618-24 MFLX97618-26 MFLX97618-27 MFLX97618-28 MFLX97618-29 MFLX97618-30 MFLX97618-31 MFLX97618-32 MFLX97618-34 MFLX97618-36 MFLX97618-38 MFLX97618-39 MFLX97618-40 MFLX97618-42 MFLX97618-44 MFLX97618-46 MFLX97618-48 MFLX97618-49
Masterflex® Ismatec® Pump Tubing, 2-Stop Microbore, Tygon® LMT-55, Avantor®
Slangar Peristaltisk pumpslang
Designed for optimal operation in your Ismatec pumps.
  • Precision extruded to tight tolerances for high repeatability, accuracy and reproducibility
  • Global market leader in range of formulations for most extensive chemical compatibility
  • Single-use custom lengths and configurations available to minimise downtime
  • Regulatory approvals to match your unique application
The overall length of LMT-55 97618-series stop tubing is 16" (406 mm). The legacy Ismatec® stop tubing was 400 mm long.
Order extension tubing under "Accessories". For technical assistanceMore Product Information