Masterflex® L/S® Precision Pump Tubing, BioPharm Platinum-Cured Silicone, Avantor®
Leverantör: Avantor Fluid Handling
MFLX96420-13EA 1675 SEK
MFLX96420-13 MFLX96420-14 MFLX96420-16 MFLX96420-17 MFLX96420-18 MFLX96420-25
Masterflex® L/S® Precision Pump Tubing, BioPharm Platinum-Cured Silicone, Avantor®
Slangar Peristaltisk pumpslang
Ultra-smooth inner surface minimizes particle entrapment.
  • Genuine Masterflex® tubing undergoes robust testing to provide optimal performance / pump life hours with peristaltic pump equipment
  • Precision extruded to tight tolerances for high repeatability, accuracy and reproducibility
  • Global market leader in range of formulations for most extensive chemical compatibility
  • Single-use custom lengths and configurations available to minimize downtime
  • Regulatory approvals to match your unique application
  • Unique packaging options include packs, cases, and spools for
More Product Information