Masterflex® L/S® 2-Stop Pump Tubing, PharMed® BPT, Avantor®
Leverantör: Avantor Fluid Handling
MFLX96114-13EA 1450 SEK
MFLX96114-13 MFLX96114-14 MFLX96114-15 MFLX96114-16 MFLX96114-24 MFLX96114-25 MFLX96114-35
Masterflex® L/S® 2-Stop Pump Tubing, PharMed® BPT, Avantor®
Slangar Peristaltisk pumpslang
Protect your light-sensitive samples.
  • Genuine Masterflex® tubing undergoes robust testing to provide optimal performance / pump life hours with peristaltic pump equipment
  • Precision extruded to tight tolerances for high repeatability, accuracy and reproducibility
  • Global market leader in range of formulations for most extensive chemical compatibility
  • Single-use custom lengths and configurations available to minimise downtime
  • Regulatory approvals to match your unique application
  • Unique packaging options include packs, cases, and spools for customer-friendly
More Product Information