Masterflex® I/P® Sanitary Fitting Precision Pump Tubing Assembly, PharMed® BPT, Avantor®
Leverantör: Avantor Fluid Handling
MFLX96102-73EA 2725 SEK
MFLX96102-73 MFLX96102-82 MFLX96103-73 MFLX96103-88 MFLX96102-70 MFLX96102-88 MFLX96203-73 MFLX96102-26 MFLX96103-26 MFLX96103-82
Masterflex® I/P® Sanitary Fitting Precision Pump Tubing Assembly, PharMed® BPT, Avantor®
Slangar Peristaltisk pumpslang
Protect your light-sensitive samples.
  • Genuine Masterflex® tubing undergoes robust testing to provide optimal performance / pump life hours with peristaltic pump equipment
  • Precision extruded to tight tolerances for high repeatability, accuracy and reproducibility
  • Global market leader in range of formulations for most extensive chemical compatibility
  • Single-use custom lengths and configurations available to minimise downtime
  • Regulatory approvals to match your unique application
Applications: Cell and tissue culture work, pharmaceutical, light-sensitiveMore Product Information