Masterflex® Ismatec® IPC High-Accuracy Multichannel Peristaltic Pumps, Avantor®
Leverantör: Avantor Fluid Handling
MFLX78006-22EA 76925 SEK
MFLX78006-22 MFLX78006-24 MFLX78006-26 MFLX78006-28 MFLX78006-42 MFLX78006-44 MFLX78006-46 MFLX78006-48 MFLX78006-22-EU MFLX78006-24-EU MFLX78006-26-EU MFLX78006-28-EU MFLX78006-42-EU MFLX78006-44-EU MFLX78006-46-EU MFLX78006-48-EU
Masterflex® Ismatec® IPC High-Accuracy Multichannel Peristaltic Pumps, Avantor®
Pumpar Slangpumpar
Achieve high-accuracy multi-channel flow, with an intuitive touch-screen interface.
  • Replicate same flow for all channels; perfect for auto-analysis, reactor studies, cell line seeding, and research
  • Planetary drive with 8 stainless steel rollers reduces pulsation, increases flow rate accuracy, and lengthens tubing life
  • 12,7 cm (5") capacitive TFT/RGB touch screen for crisp resolution and a wide viewing angle
  • Flexible operational control via screen or remote control (analog remote control, serial, or EtherNet/IP)
  • Three-level user access – gives you the
More Product Information