Masterflex® L/S® Standard Digital Pump Systems, Avantor®
Leverantör: Avantor Fluid Handling
MFLX77919-25-EUEA 70050 SEK
MFLX77919-25-EU MFLX77919-35-EU MFLX77921-65-EU MFLX77921-75-EU MFLX77921-85-EU
Masterflex® L/S® Standard Digital Pump Systems, Avantor®
Pumpar Slangpumpar
Designed with maintenance-free high-accuracy motor and a graphical interface—makes setup and operation easier than ever!
  • Digital interface with graphical LCD shows pump performance and operating modes—continuous run, timed dispense, copy dispense, or volume dispense
  • Select your preferred language from the user-friendly menu for programming and operation
  • Dispense by Volume in ml (0.001 to 9,999 mL), L, or gal; Copy—1 to 9,999 dispense cycles; or Time—time interval of on/off cycles (1 sec to 99 hr, 59 min, 59 sec per cycle)Antidrip function ensures
More Product Information