Masterflex® Interchangeable Power Cord Assemblies - IEC to Unique Country Plug, Avantor®
Leverantör: Avantor Fluid Handling
MFLX50001-70EA 880 SEK
MFLX50001-70
Masterflex® Interchangeable Power Cord Assemblies - IEC to Unique Country Plug, Avantor®
Kablar
Optimize performance of your Masterflex® pump.
  • Choose from a wide range of power cords
Choose an interchangeable cord/plug assembly for 115 or 230 VAC operation. Each 6-ft (1,8 m) cord and moded plug assembly features an IEC 320 appliance connector and rated for 10 A at 250 VAC.