Value Plastics® Adapter Fittings, Male Luer to Threaded, Straight
Leverantör: Avantor Fluid Handling
MFLX45518-82BLKEA 4550 SEK
MFLX45518-82BLK MFLX45518-88BLK
Value Plastics® Adapter Fittings, Male Luer to Threaded, Straight
Beslag och kopplingar
Easily transition between luer fittings and UNF or NPT threads.
  • Ideal for general-purpose and single-use applications, to connect tubing with rest of system
These adapters transition between luer fittings and UNF or NPT threads. Both male and female luer connections are available. All threaded connections are male thread.