Masterflex® Adapter Fittings, Luer to Threaded, Avantor®
Leverantör: Avantor Fluid Handling
MFLX45502-60EA 290 SEK
MFLX45502-60 MFLX45502-66 MFLX45505-76 MFLX45505-78 MFLX45505-80 MFLX45505-82 MFLX45505-84 MFLX45505-86 MFLX45505-88
Masterflex® Adapter Fittings, Luer to Threaded, Avantor®
Beslag och kopplingar
Easily transition to a luer fitting.
  • Repackaged in an ISO 9001 facility in an ISO Class 7 Cleanroom
  • Ideal for general-purpose and single-use applications, to connect tubing with rest of system
  • Determine which connector type is needed for your application by contacting free technical application assistance*
  • Global market leader in range of materials for most extensive chemical compatibility
  • Single-use custom configuration solutions to minimise downtime
  • Regulatory approvals to match your unique application
  • Unique packaging options include individuals, packs,
More Product Information