Liquid-Solids Separator / Concentrator Units
Leverantör: Avantor Fluid Handling
29995-02 29995-06 29995-12 29995-18
MFLX29995-02EA 10950 SEK
MFLX29995-02 MFLX29995-06 MFLX29995-12 MFLX29995-18
Liquid-Solids Separator / Concentrator Units
Pumpar Centrifugalavskiljare
Proven design utilizes centrifugal action to remove and concentrate solids.
  • No screens, cartridges or filters to clean or replace
  • Backflushing is not required - minimizing the loss of water and process fluids
  • Unit has no moving parts - less maintenance down time required
In many processes, removal of solids can extend the usefulness of the process media and protect equipment from fouling and abrasive wear; regulatory benefits also exist in many applications. The basic design requires no electrical power and minimal piping, which makes cost payback onMore Product Information