Masterflex® L/S® Digital Console Process Drive Replacement Seal Kit, Avantor®
Leverantör: Avantor Fluid Handling
MFLX07575-01EA 4600 SEK
MFLX07575-01
Masterflex® L/S® Digital Console Process Drive Replacement Seal Kit, Avantor®
Pumpar Slangpumpar
Make the most of your Masterflex drives replacement parts and other accessories.
  • Suitable for use with digital drive 07575-10
Extend the life of your Masterflex pump with these replacement parts.
Leveransinformation: Includes shaft seal, hardware, and gasket.