Masterflex® Ismatec® Pump Tubing, Peroxide-Cured Silicone, Avantor®
Leverantör: Avantor Fluid Handling
MFLX06516-34EA 1775 SEK
MFLX06516-34
Masterflex® Ismatec® Pump Tubing, Peroxide-Cured Silicone, Avantor®
Slangar Peristaltisk pumpslang
Designed for optimal operation in your Ismatec pumps.
  • Precision extruded to tight tolerances for high repeatability, accuracy and reproducibility
  • Global market leader in range of formulations for most extensive chemical compatibility
  • Single-use custom lengths and configurations available to minimise downtime
  • Regulatory approvals to match your unique application
Silicone tubing blended with organic peroxide for biological applications. This economical tubing has long life, making it ideal for lab and general applications.