Masterflex® Transfer Tubing, C-Flex®, Clear, Avantor®
Leverantör: Avantor Fluid Handling
MFLX06422-00EA 535 SEK
MFLX06422-00 MFLX06422-01 MFLX06422-02 MFLX06422-03 MFLX06422-04 MFLX06422-05 MFLX06422-06 MFLX06422-07 MFLX06422-08 MFLX06422-09 MFLX06422-10 MFLX06422-11 MFLX06422-12 MFLX06422-13 MFLX06422-14 MFLX06422-15 MFLX06422-16 MFLX06422-17 MFLX06422-18 MFLX06422-21 MFLX06422-23 MFLX06424-95 MFLX06424-96
Masterflex® Transfer Tubing, C-Flex®, Clear, Avantor®
Slangar Överföringsslang
Meets the demands of various industries.
  • Ideal for general-purpose, single-use, and extended fluid path applications
  • Global market leader in range of formulations for most extensive chemical compatibility
  • Single-use custom lengths and configurations available to minimise downtime
  • Regulatory approvals to match your unique application
Applications: Pharmaceutical, biotech, and general-purpose lab uses requiring high performance. Economical alternative to platinum silicone.
Characteristics: Exceptional tensile and tear strength, good biocompatibility,More Product Information