Masterflex® Ismatec® Microbore Pump Tubing, Tygon® LMT-55, Avantor®
Leverantör: Avantor Fluid Handling
MFLX05502-10EA 1125 SEK
MFLX05502-10 MFLX05502-12 MFLX05502-14 MFLX05502-16 MFLX05502-18 MFLX05502-19 MFLX05502-22 MFLX05502-24 MFLX05502-26 MFLX05502-27 MFLX05502-28 MFLX05502-30 MFLX05502-31 MFLX05502-32 MFLX05502-34 MFLX05502-36 MFLX05502-38 MFLX05502-39 MFLX05502-40 MFLX05502-42 MFLX05502-44 MFLX05502-46 MFLX05502-48 MFLX05502-49 MFLX05502-09
Masterflex® Ismatec® Microbore Pump Tubing, Tygon® LMT-55, Avantor®
Slangar Peristaltisk pumpslang
Crystal clear and flexible while also being long-lasting and crack-resistant.
  • Tubing undergoes robust testing to provide optimal performance / pump life hours with peristaltic pump equipment
  • Precision extruded to tight tolerances for high repeatability, accuracy and reproducibility
  • Global market leader in range of formulations for most extensive chemical compatibility
  • Regulatory approvals to match your unique application
Designed for optimal operation in your Ismatec pumps.
Applications: General laboratory, food and beverage, biopharmaceutical,More Product Information