Tubing, FEP
Leverantör: METROHM
6.1805.010 6.1805.030 6.1805.110 6.1805.080 6.1805.100 6.1829.020 6.1805.460
METR6.1805.010EA 1525 SEK
METR6.1805.010 METR6.1805.030 METR6.1805.110 METR6.1805.080 METR6.1805.100 METR6.1829.020 METR6.1805.460
Tubing, FEP
Slangar Överföringsslang
FEP, translucent.
  • With light and kink protection
  • With thread connections at both ends
Material allows much longer lengths and tighter tolerances than PTFE tubing.