1,1,1,3,3,3-Hexafluoro-2-propanol zS

Leverantör: Merck
Fara

Synonyms: 1,1,1,3,3,3-Hexafluoroisopropyl alcohol, Hexafluoro-2-propanol, Hexafluoroisopropanol, 1,1,1,3,3,3-Hexafluoropropan-2-ol, HFIP

MQ200

8.04515.0025 8.04515.0100 8.04515.1000
MERC8.04515.0025EA 870 SEK
MERC8.04515.0025 MERC8.04515.0100 MERC8.04515.1000
1,1,1,3,3,3-Hexafluoro-2-propanol zS
1,1,1,3,3,3-Hexafluoro-2-propanol
Formel: C₃H₂F₆O
MW: 168.04 g/mol
Kokpunkt: 59.1
Smältpunkt: –96 °C
Densitet: 1.492 g/cm³ (20 °C)
Flampunkt: >100 °C
Storage Temperature: Omgivande
MDL Number: MFCD00011651
CAS nummer: 920-66-1
EINECS: 213-059-4
UN: 3265
ADR: 8 II
Merck Index: 13,09598

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR