2-Bromo-2'-chloroacetanilide

Leverantör: Thermo Scientific

Synonyms: N-(Bromoacetyl)-2-chloroaniline

SPB05477DA SPB05477EA
MAYBSPB05477DAEA 0 SEK
MAYBSPB05477DA MAYBSPB05477EA
2-Bromo-2'-chloroacetanilide
2-Bromo-2'-chloroacetanilide
Formel: C₈H₇BrClNO
MDL Number: MFCD00173815
CAS nummer: 5439-11-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR