2,4-Dichlorobenzaldehyde oxime

Leverantör: Thermo Scientific

SPB04865DA SPB04865EA
MAYBSPB04865DAEA 0 SEK
MAYBSPB04865DA MAYBSPB04865EA
2,4-Dichlorobenzaldehyde oxime
2,4-Dichlorobenzaldehyde oxime
Formel: C₇H₅Cl₂NO
MW: 190.03
CAS nummer: 56843-28-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR