1,3-Dimethyl-1H-pyrazole-5-carbonyl chloride

Leverantör: Thermo Scientific

CC12502CB CC12502DA
MAYBCC12502CBEA 0 SEK
MAYBCC12502CB MAYBCC12502DA
1,3-Dimethyl-1H-pyrazole-5-carbonyl chloride
1,3-Dimethyl-1H-pyrazole-5-carbonyl chloride
Formel: C₆H₇ClN₂O
MDL Number: MFCD00833258
CAS nummer: 55458-67-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR