N-[2-Nitro-4-(trifluoromethyl)phenyl]piperazine

Leverantör: Thermo Scientific

BTB03244DA BTB03244EA
MAYBBTB03244DAEA 0 SEK
MAYBBTB03244DA MAYBBTB03244EA
N-[2-Nitro-4-(trifluoromethyl)phenyl]piperazine
N-[2-Nitro-4-(trifluoromethyl)phenyl]piperazine
Formel: C₁₁H₁₂F₃N₃O₂
MDL Number: MFCD00052613
CAS nummer: 58315-38-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR