WHEATON® Autoclave Bags, High Temperature
Leverantör: DWK Life Sciences
370100 370110 370120
LSUK370100EA 1075 SEK
LSUK370100 LSUK370110 LSUK370120
WHEATON® Autoclave Bags, High Temperature
Påsar Steriliseringspåsar
WHEATON® autoclave bags are available in three different sizes and are printed with blue hazard warnings labels for contaminated waste disposal.
  • Manufactured from strong polypropylene
  • Ideal for the disposal of all biological waste in autoclaves or incinerators
  • Uniform wall thickness and seamless bottom, designed to ensure high pressure bearing
Suitable for use within the two defined high temperature sterilisation methods; autoclave at 121°C for 30 minutes or autoclave at 134°C for 10 minutes.
All bags are printed with blue bio-hazard labelling
More Product Information