Mueller Hinton Fastidious Agar
Leverantör: LIOFILCHEM
10132
LIOF10132EA 550 SEK
LIOF10132
Mueller Hinton Fastidious Agar
Media Dehydrerade mikrobiologimedia
Mueller Hinton fastidious agar (Horse blood 5% + 20 mg/L β-NAD) is recommended by EUCAST for antimicrobial susceptibility testing of Streptococcus spp (including S. pneumoniae), Haemophilus spp and several other fastidious organisms.