Flexible couplings for stirrers
Leverantör: Lenz Laborglas GmbH & CO.KG
06400000
LENZ06400000EA 3250 SEK
LENZ06400000
Flexible couplings for stirrers
Omrörare Omrörningsverktyg
Flexible coupling made of stainless steel.
  • Compensates for axial and height differences
  • No adjustments necessary for motor and stirring rod
  • Stainless steel clamping chuck with clamping range 8 to 10 mm