ALiCE for Research

Leverantör: LENIOBIO

AL00000002 AL00000003
LENOAL00000002EA 11050 SEK
LENOAL00000002 LENOAL00000003
ALiCE for Research
Proteinexpressions Reagens för proteinexpression

From ALiCE® for Research to ALiCE® for Scale-Up Protein Expression, we offer cell-free protein expression products with radical results for every production setup.


  • Produces a minimum of 2 mg/ml protein in under 48 hours
  • Simple reaction set-up - just add DNA
  • Capable of catalysing complex post-translational modifications


The ALiCE® kits were developed specifically for ease of use. Expression of your protein of interest follows a simple four-step procedure:
1. Clone your gene of interest into the ALiCE® vector of your choosing
2. Add your plasmid DNA to a thawed vial of ALiCE® lysate
3. Incubate at 25 °C with aeration for 24 - 48 h
4. Collect your protein
For the full details of the procedure, download our user manual.


ALiCE® for Research-Mini kit contains reagents for 6×50 µl ALiCE protein expression reactions:
6×50 µl ALiCE lysate aliquots
pALiCE01 vector
pALiCE02 vector
6×ALiCE reaction tubes
Short protocol


ALiCE® for Research-Midi kit contains regents for protein cell free expression reactions:
6×200 µl ALiCE lysate aliquots
pALiCE01 vector
pALiCE02 vector
24×ALiCE reaction tubes
Short protocol


ALiCE® for Research-Mixi kit contains reagents for 60×500 µl ALiCE protein expression reactions:
6×500 µl ALiCE lysate aliquots
pALiCE01 vector
pALiCE02 vector
60×ALiCE reaction tubes
Short protocol


Storage Conditions: They should be transferred directly to a –80 °C freezer upon delivery.


Beställningsinformation: Temp data logger can be added additionally.


Förpackning: Dry Ice shipping.


Observera: The ALiCE® Cell-free protein expression kits are intended for research use only.

New Product

Order Now

SPECIFIKATIONER

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR