Balance enclosures, XPert™
Leverantör: LABCONCO
3930221 3930321 3930421 3930230 3930241 3930242 3930300 3930322 3930330 3930342 3930400 3930422 3930430 3930441 3930442 3971500 3971521 3971522 3971530 3971541 3971542 3971560 3971571 3971572 3971600 3971621 3971622 3971630 3971641 3971642 3971660 3971671 3971672 3971800 3971821 3971822 3971830 3971841 3971842 3971860 3971871 3971872 3930341
LBCP3930221EA 0 SEK
LBCP3930221 LBCP3930321 LBCP3930421 LBCP3930230 LBCP3930241 LBCP3930242 LBCP3930300 LBCP3930322 LBCP3930330 LBCP3930342 LBCP3930400 LBCP3930422 LBCP3930430 LBCP3930441 LBCP3930442 LBCP3971500 LBCP3971521 LBCP3971522 LBCP3971530 LBCP3971541 LBCP3971542 LBCP3971560 LBCP3971571 LBCP3971572 LBCP3971600 LBCP3971621 LBCP3971622 LBCP3971630 LBCP3971641 LBCP3971642 LBCP3971660 LBCP3971671 LBCP3971672 LBCP3971800 LBCP3971821 LBCP3971822 LBCP3971830 LBCP3971841 LBCP3971842 LBCP3971860 LBCP3971871 LBCP3971872 LBCP3930341
Balance enclosures, XPert™
Dragskåp Extraktionsskåp
XPert™ balance enclosures provide user protection by keeping powders, particulates and fume contained during weighing procedures. They are well suited for pharmaceutical research and other laboratory applications involving weighing of toxic substances. These vented balance safety enclosures may be ducted to the outside or connected to the FilterMate Portable Exhauster to be exhausted back into the laboratory through HEPA filters, carbon filters, or a combination of both HEPA and carbon filters.
  • The deep interior easily accommodates large micro and analytical balances
  • Clean-Sweep™
More Product Information