Balance enclosures, XPert™ Easy-order
Leverantör: LABCONCO
3960302 3960421 3960221 3960222 3960321 3960322 3960422
LBCP3960302EA 0 SEK
LBCP3960302 LBCP3960421 LBCP3960221 LBCP3960222 LBCP3960321 LBCP3960322 LBCP3960422
Balance enclosures, XPert™ Easy-order
Dragskåp Extraktionsskåp
XPert™ balance enclosures provide user protection by keeping powders, particulates and fume contained during weighing procedures. They are well suited for pharmaceutical research and other laboratory applications involving weighing of toxic substances. These vented balance safety enclosures may be ducted to the outside or connected to the FilterMate Portable Exhauster to be exhausted back into the laboratory through HEPA filters.
  • The deep interior easily accommodates large micro and analytical balances
  • Clean-Sweep air foil allows air to sweep the work
More Product Information